Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193240.2 | NP_001180169.1 | 5 - 6 | 772 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |