Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014844.3 | NP_001014844.1 | 2 - 3 | 121 | 61 | |
GenBank mRNA | BC102942.1 | - | 2 - 3 | 122 | 61 | |
BT021526.1 | - | 1 - 2 | 74 | 61 | ||
BT021895.1 | - | 1 - 2 | 68 | 61 | ||
BT025344.1 | - | 1 - 2 | 62 | 61 | ||
BT026160.1 | - | 1 - 2 | 63 | 61 | ||
BT026274.1 | - | 1 - 2 | 62 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |