Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033620.2 | NP_001028792.1 | 1 - 2 | 246 | 60 | |
GenBank mRNA | AJ510149.1 | - | 1 - 2 | 141 | 60 | |
BC102141.1 | - | 1 - 2 | 247 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |