Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035099.1 | NP_001030271.1 | 2 - 3 | 408 | 89 | |
GenBank mRNA | BC102513.1 | - | 2 - 3 | 408 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |