Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035099.1 | NP_001030271.1 | 6 - 7 | 780 | 82 | |
GenBank mRNA | BC102513.1 | - | 6 - 7 | 780 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |