Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205376.2 | NP_001192305.1 | 3 - 4 | 168 | 101 | |
GenBank mRNA | BC103023.1 | - | 3 - 4 | 159 | 101 | |
EH182364.1 | - | 3 - 4 | 168 | 101 | ||
EV683183.1 | - | 3 - 4 | 168 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |