Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130764.1 | NP_001124236.1 | 2 - 3 | 1949 | 68 | |
GenBank mRNA | L27218.1 | - | 2 - 3 | 1949 | 68 | |
S69583.1 | - | 2 - 3 | 1949 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |