Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270317.4 | NP_001257246.2 | 16 - 17 | 3908 | 125 | |
NM_001373583.1 | NP_001360775.1 | 16 - 17 | 3875 | 125 | ||
NM_001381151.1 | NP_001368758.1 | 16 - 17 | 3891 | 125 | ||
NM_001381152.1 | NP_001368759.1 | 16 - 17 | 3891 | 125 | ||
NM_001381153.1 | NP_001368756.1 | 16 - 17 | 3858 | 125 | ||
NM_001381154.1 | NP_001368757.1 | 16 - 17 | 3858 | 125 | ||
NM_001381155.1 | NP_001368146.1 | 16 - 17 | 3885 | 125 | ||
NM_001381156.2 | NP_001368760.1 | 16 - 17 | 3881 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |