Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322663.4 | NP_001309674.1 | 15 - 16 | 2249 | 64 | |
NM_001322664.3 | NP_001309467.1 | 14 - 15 | 2210 | 64 | ||
NM_001322665.4 | NP_001309678.1 | 10 - 11 | 1321 | 64 | ||
NM_001322667.4 | NP_001309677.1 | 4 - 5 | 784 | 64 | ||
NM_001322668.4 | NP_001309676.1 | 4 - 5 | 697 | 64 | ||
NM_001383675.1 | 14 - 15 | 2203 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |