Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078107.6 | NP_510508.1 | 2 - 3 | 268 | 106 | |
GenBank mRNA | AB012700.1 | - | 2 - 3 | 290 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |