Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078107.6 | NP_510508.1 | 5 - 6 | 573 | 82 | |
GenBank mRNA | AB012700.1 | - | 5 - 6 | 595 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |