Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029782.5 | NP_001024953.1 | 4 - 5 | 459 | 114 | |
GenBank mRNA | AY349017.1 | - | 4 - 5 | 481 | 114 | |
AY349018.1 | - | 1 - 2 | 121 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |