Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olrn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029163.7 | NP_001024334.1 | 4 - 5 | 1308 | 107 | |
NM_001373597.2 | NP_001359691.1 | 4 - 5 | 1294 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |