Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipi-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078172.9 | NP_510573.3 | 3 - 4 | 1137 | 78 | |
GenBank mRNA | EU043523.1 | - | 3 - 4 | 1134 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |