Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078187.7 | NP_510588.1 | 4 - 5 | 925 | 74 | |
GenBank mRNA | U10411.1 | - | 4 - 5 | 947 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |