Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029682.5 | NP_001024853.1 | 4 - 5 | 475 | 79 | |
NM_001029684.6 | NP_001024855.1 | 3 - 4 | 372 | 79 | ||
NM_001381184.1 | NP_001367802.1 | 2 - 3 | 280 | 79 | ||
GenBank mRNA | AY204163.1 | - | 3 - 4 | 356 | 79 | |
AY204164.1 | - | 3 - 4 | 370 | 79 | ||
AY204165.1 | - | 3 - 4 | 370 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |