Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gap-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306410.1 | NP_001293339.1 | 2 - 3 | 211 | 85 | |
NM_058515.6 | NP_490916.5 | 9 - 10 | 1250 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |