Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374886.2 | NP_001361963.1 | 13 - 14 | 3928 | 115 | |
NM_001380120.2 | NP_001367609.1 | 13 - 14 | 3943 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |