Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tom-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129022.5 | NP_001122494.1 | 23 - 24 | 3349 | 97 | |
NM_001129023.2 | NP_001122495.1 | 23 - 24 | 3367 | 97 | ||
NM_001313498.3 | NP_001300427.1 | 23 - 24 | 3716 | 97 | ||
NM_001313499.3 | NP_001300428.1 | 22 - 23 | 3704 | 97 | ||
NM_001313500.3 | NP_001300429.1 | 23 | 3624 | 97 | ||
NM_059222.7 | NP_491623.3 | 22 - 23 | 3338 | 97 | ||
NM_059225.5 | NP_491626.2 | 5 - 6 | 455 | 97 | ||
GenBank mRNA | AY912102.1 | - | 22 - 23 | 3231 | 97 | |
AY997305.1 | - | 23 - 24 | 3243 | 97 | ||
AY997306.1 | - | 23 - 24 | 3258 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |