Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026412.4 | NP_001021583.1 | 2 - 3 | 274 | 75 | |
GenBank mRNA | AB040992.1 | - | 2 - 3 | 279 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |