Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026601.6 | NP_001021772.1 | 1 - 2 | 158 | 115 | |
NM_001383734.1 | 2 - 3 | 293 | 115 | |||
NM_170927.5 | NP_740922.1 | 1 - 2 | 158 | 115 | ||
GenBank mRNA | AF332568.1 | - | 1 - 2 | 152 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |