Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bbs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170937.3 | NP_740933.1 | 3 - 4 | 621 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |