Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osta-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171004.7 | NP_741003.1 | 2 - 3 | 278 | 117 | |
NR_136362.1 | 2 - 3 | 278 | 117 | |||
NR_136363.1 | 2 - 3 | 278 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |