Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cam-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026736.5 | NP_001021907.1 | 8 - 9 | 2640 | 94 | |
NM_001026737.5 | NP_001021908.1 | 8 - 9 | 2672 | 94 | ||
NM_001383915.1 | 8 - 9 | 2635 | 94 | |||
GenBank mRNA | AJ132946.1 | - | 8 - 9 | 2639 | 94 | |
AJ132947.1 | - | 7 - 8 | 2560 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |