Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171029.8 | NP_741034.2 | 7 - 8 | 1294 | 106 | |
NM_171855.4 | NP_741035.2 | 8 - 9 | 1474 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |