Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383939.1 | 1 - 2 | 126 | 130 | ||
NM_001383940.1 | 1 - 2 | 120 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |