Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171035.6 | NP_741041.1 | 3 - 4 | 423 | 83 | |
GenBank mRNA | LC026472.1 | - | 2 - 3 | 367 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |