Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026998.5 | NP_001022169.1 | 17 - 18 | 2471 | 65 | |
NM_001026999.8 | NP_001022170.1 | 19 - 20 | 2807 | 65 | ||
NM_001027000.6 | NP_001022171.1 | 16 - 17 | 2602 | 65 | ||
NM_001027001.5 | NP_001022172.1 | 11 - 12 | 1547 | 65 | ||
NM_001267342.3 | NP_001254271.1 | 17 - 18 | 2471 | 65 | ||
NM_001267343.3 | NP_001254272.1 | 18 - 19 | 2684 | 65 | ||
NM_001267344.3 | NP_001254273.1 | 12 - 13 | 1883 | 65 | ||
NM_171039.3 | NP_741045.1 | 18 - 19 | 2635 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |