Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026998.5 | NP_001022169.1 | 9 - 10 | 1279 | 91 | |
NM_001026999.8 | NP_001022170.1 | 9 - 10 | 1279 | 91 | ||
NM_001027000.6 | NP_001022171.1 | 6 - 7 | 1074 | 91 | ||
NM_001267342.3 | NP_001254271.1 | 9 - 10 | 1279 | 91 | ||
NM_001267343.3 | NP_001254272.1 | 9 - 10 | 1279 | 91 | ||
NM_001267344.3 | NP_001254273.1 | 2 - 3 | 355 | 91 | ||
NM_171039.3 | NP_741045.1 | 9 - 10 | 1320 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |