Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zmp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171138.3 | NP_741156.1 | 2 - 3 | 307 | 74 | |
GenBank mRNA | AB007817.1 | - | 2 - 3 | 307 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |