Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exos-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372617.1 | NP_001360028.1 | 1 - 2 | 65 | 77 | |
NM_171186.6 | NP_741217.1 | 2 - 3 | 91 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |