Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027810.5 | NP_001022981.1 | 12 - 13 | 2354 | 76 | |
NM_001129275.4 | NP_001122747.1 | 2 - 3 | 457 | 76 | ||
NM_001129276.3 | NP_001122748.1 | 12 - 13 | 2354 | 76 | ||
NM_001268079.4 | NP_001255008.1 | 13 - 14 | 2453 | 76 | ||
NM_001268080.3 | NP_001255009.1 | 8 - 9 | 1543 | 76 | ||
NM_001306679.3 | NP_001293608.1 | 6 - 7 | 1339 | 76 | ||
NM_001306687.3 | NP_001293616.1 | 7 - 8 | 1357 | 76 | ||
NM_001306688.3 | NP_001293617.1 | 2 - 3 | 457 | 76 | ||
NM_001382965.1 | 14 - 15 | 2675 | 76 | |||
NM_171221.7 | NP_741263.3 | 14 - 15 | 2445 | 76 | ||
GenBank mRNA | AF424978.1 | - | 12 - 13 | 2290 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |