Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380163.1 | NP_001367796.1 | 7 - 8 | 1524 | 120 | |
NM_001380164.2 | NP_001367795.1 | 7 - 8 | 1538 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |