Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171311.6 | NP_741365.1 | 4 - 5 | 390 | 90 | |
NM_171312.5 | NP_741366.1 | 3 - 4 | 367 | 90 | ||
NM_171903.6 | NP_741367.1 | 3 - 4 | 304 | 90 | ||
NR_131466.1 | 3 - 4 | 247 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |