Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026099.5 | NP_001021270.1 | 10 - 11 | 1186 | 113 | |
NM_001383114.1 | 10 - 11 | 1193 | 113 | |||
NR_131561.1 | 10 - 11 | 1150 | 113 | |||
GenBank mRNA | AB024085.1 | - | 10 - 11 | 1193 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |