Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026099.5 | NP_001021270.1 | 5 - 6 | 567 | 102 | |
NM_001383114.1 | 5 - 6 | 574 | 102 | |||
NR_131561.1 | 5 - 6 | 531 | 102 | |||
GenBank mRNA | AB024085.1 | - | 5 - 6 | 574 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |