Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027908.6 | NP_001023079.1 | 11 - 12 | 4632 | 77 | |
NM_171379.6 | NP_741442.1 | 11 - 12 | 4632 | 77 | ||
GenBank mRNA | AF023602.1 | - | 12 - 13 | 4665 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |