Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bli-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026369.6 | NP_001021540.1 | 12 - 13 | 2123 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Caenorhabditis elegans Probe spans exons |