Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: knl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380547.1 | NP_001367697.1 | 2 - 3 | 278 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |