Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361754.3 | NP_001348668.1 | 4 - 5 | 1271 | 137 | |
NM_059465.5 | NP_491866.2 | 4 - 5 | 1272 | 137 | ||
GenBank mRNA | AY887907.1 | - | 4 - 5 | 1288 | 137 | |
AY954523.1 | - | 4 - 5 | 1288 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |