Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tre-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059489.7 | NP_491890.2 | 3 - 4 | 527 | 125 | |
GenBank mRNA | AJ512337.1 | - | 3 - 4 | 355 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |