Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025858.4 | NP_001021029.1 | 7 - 8 | 1471 | 67 | |
NM_001306351.3 | NP_001293280.1 | 5 - 6 | 809 | 67 | ||
NM_001306352.3 | NP_001293281.1 | 3 - 4 | 362 | 67 | ||
NM_059539.6 | NP_491940.2 | 6 - 7 | 1284 | 67 | ||
GenBank mRNA | AF170425.1 | - | 7 - 8 | 1240 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |