Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cye-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025856.4 | NP_001021027.1 | 3 - 4 | 308 | 93 | |
GenBank mRNA | AF058331.1 | - | 3 - 4 | 328 | 93 | |
AF520616.1 | - | 3 - 4 | 330 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |