Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263462.4 | NP_001250391.1 | 5 - 6 | 1490 | 105 | |
NM_001263463.4 | NP_001250392.1 | 4 - 5 | 1358 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |