Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059617.6 | NP_492018.1 | 8 - 9 | 1880 | 92 | |
GenBank mRNA | AB000913.1 | - | 8 - 9 | 1898 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |