Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crml-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322686.3 | NP_001309450.1 | 5 - 6 | 960 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |