Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: goa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059707.6 | NP_492108.1 | 5 - 6 | 821 | 72 | |
GenBank mRNA | AY008140.1 | - | 5 - 6 | 727 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |