Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263626.3 | NP_001250555.1 | 8 - 9 | 1853 | 85 | |
NM_001263627.3 | NP_001250556.1 | 7 - 8 | 1600 | 85 | ||
GenBank mRNA | AF159144.1 | - | 8 - 9 | 1853 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |