Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ego-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059731.6 | NP_492132.1 | 13 - 14 | 4423 | 93 | |
GenBank mRNA | AF159143.1 | - | 13 - 14 | 4421 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |