Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rskn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047180.4 | NP_001040645.1 | 1 - 2 | 224 | 99 | |
NM_001361775.4 | NP_001348666.1 | 1 - 2 | 226 | 99 | ||
NM_001361776.3 | NP_001348667.1 | 1 - 2 | 224 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |